Learn More
Wilmad Labglass PIPET VOL CLASS B 1ML
Supplier:  Wilmad Labglass LG9355102
Encompass_Preferred
Volumetric Pipet, Class 'B', To Deliver, Color-Coded. Capacity: 1mL. Capacity Tolerance: 0.012mL. Color Band: Blue. Manufactured to meet the requirements for accuracy, appearance, glass quality and calibration line accuracy of ASTM E969.
Catalog No. 50-807-923
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product