Learn More
Wilmad Labglass PIPET VOL CLASS B 2ML
Supplier:  Wilmad Labglass LG9355104
Encompass_Preferred
Volumetric Pipet, Class 'B', To Deliver, Color-Coded. Capacity: 2mL. Capacity Tolerance: 0.012mL. Color Band: Orange. Manufactured to meet the requirements for accuracy, appearance, glass quality and calibration line accuracy of ASTM E969.
Catalog No. 50-807-924