Learn More
Wilmad Labglass PIPET VOL CLASS B 5ML
Supplier:  Wilmad Labglass LG9355110
Encompass_Preferred
Volumetric Pipet, Class 'B', To Deliver, Color-Coded. Capacity: 5mL. Capacity Tolerance: 0.02mL. Color Band: White. Manufactured to meet the requirements for accuracy, appearance, glass quality and calibration line accuracy of ASTM E969.
Catalog No. 50-802-848