Learn More
Hach Company Pipet-Volumetric-0.5 mL
Supplier:  Hach Company 1451534
Encompass
Pipet-Volumetric-Class A-0 5 mLGlass Class A pipets unserialized and calibrated To Deliver Designed to ASTM E969 specification for accuracy and ASTM E1273 for color coding
Catalog No. 50-002-57151