Learn More
Hach Company Pipet, Volumetric, Class A, 10 mL
Supplier:  Hach Company 1451538
Encompass
Glass Class A pipets unserialized and calibrated “To Deliver”. Designed to ASTM E969 specification for accuracy and ASTM E1273 for color coding.
Catalog No. 50-199-5054