Learn More
Integra Biosciences Corp PIPETBOY acu 2 pipet aid, Blue
SDP
Catalog No. NC0085685
Encompass
The second generation of PIPETBOY acu pipet aid is equipped with the latest technology providing an increased pipetting speed and a significant weight reduction of the instrument. These two features help you to cut your work time and offer you an improved ergonomic pipetting experience. Besides these changes the main benefits of PIPETBOY ACU were preserved: the unmatched pipetting precision, the colorful look and the weight balance. Autoclavable nose piece and easy to replace hydrophobic filter for reliable protection. Sensitive control of the liquid by applying various finger pressure on triggers, from fine to fast pipetting.
Catalog No. NC0085685 Supplier Integra Biosciences Corp Supplier No. 155017
The second generation of PIPETBOY acu pipet aid is equipped with the latest technology providing an increased pipetting speed and a significant weight reduction of the instrument. These two features help you to cut your work time and offer you an improved ergonomic pipetting experience. Besides these changes the main benefits of PIPETBOY ACU were preserved: the unmatched pipetting precision, the colorful look and the weight balance. Autoclavable nose piece and easy to replace hydrophobic filter for reliable protection. Sensitive control of the liquid by applying various finger pressure on triggers, from fine to fast pipetting.