Learn More
Integra Biosciences Corp PIPETBOY PRO GREEN
SDP
Catalog No. NC9062813
Encompass
Liquid handling lies at the core of every life science experiment and a key to successful research is the ability to handle liquids with precision and efficiency. The PIPETBOY pro pipet aid unites all essential characteristics for productive work with sero-logical pipettes: accuracy, speed and ergonomic design. The accuracy of pipetting depends on precise graduations on the serological pipette and on the ability to accurately match the liquid's meniscus with the graduation mark. The PIPETBOY pro enables the user to very accurately displace liquid in the pipette by applying variable finger pressure on the pipettor triggers. Maximum pipetting speed: 12 ml/s; Presetting of speed (via thumb wheel): 2 -
Catalog No. NC9062813 Supplier Integra Biosciences Corp Supplier No. 156402
Liquid handling lies at the core of every life science experiment and a key to successful research is the ability to handle liquids with precision and efficiency. The PIPETBOY pro pipet aid unites all essential characteristics for productive work with sero-logical pipettes: accuracy, speed and ergonomic design. The accuracy of pipetting depends on precise graduations on the serological pipette and on the ability to accurately match the liquid's meniscus with the graduation mark. The PIPETBOY pro enables the user to very accurately displace liquid in the pipette by applying variable finger pressure on the pipettor triggers. Maximum pipetting speed: 12 ml/s; Presetting of speed (via thumb wheel): 2 -