Learn More
Cole-Parmer Essentials Standard-Length Serological Pipette, 50 mL, Ind. Pack; 90/CS
Catalog No. NC1888984 Shop All Cole-Parmer Products
Encompass_Preferred
Easily recognize pipette capacity—color-coded stripes and matching packagingVirgin polystyrene is gamma irradiated for sterilitySterile pipettes are available bulk bagged, or individually wrapped and then bulk packed in bagsRead while filling or dispensing—features bold-colored positive and negative graduationsMaterial plug protects your pipette controller from biocontamination and harmful vapors⌡Individually wrapped in plastic with paper-backed packages, then bulk packed in bags of 30; 3 bags per caseStandard-length serological pipettes—manufactured to ASTM standard E-934 for serological pipettes.
Catalog No. NC1888984 Supplier Cole-Parmer Supplier No. 1300260
Easily recognize pipette capacity—color-coded stripes and matching packagingVirgin polystyrene is gamma irradiated for sterilitySterile pipettes are available bulk bagged, or individually wrapped and then bulk packed in bagsRead while filling or dispensing—features bold-colored positive and negative graduationsMaterial plug protects your pipette controller from biocontamination and harmful vapors⌡Individually wrapped in plastic with paper-backed packages, then bulk packed in bags of 30; 3 bags per caseStandard-length serological pipettes—manufactured to ASTM standard E-934 for serological pipettes.