Learn More
Electron Microscopy Sciences Graduated Transfer Pipettes 4.8 ml, 1.8 ml Bulb draw, Sterile individual package
SDP
Catalog No. 501899396
Encompass_Preferred
These polyethylene disposable have a low-affinity surface, attached bulb avoids cross-contamination, and are one step biohazard disposable. These pipets are ideal for clinical applications, blood banking, remove, disperse small volume, transfer in and out of multi well plates, reagent handing, pH meter maintenance, etc.
Bulb Draw: 1.8 ml
Drops/mL: 24
OAL: 15.2 cm
Catalog No. 50-189-9396 Supplier Electron Microscopy Sciences Supplier No. 7096210S
These polyethylene disposable have a low-affinity surface, attached bulb avoids cross-contamination, and are one step biohazard disposable. These pipets are ideal for clinical applications, blood banking, remove, disperse small volume, transfer in and out of multi well plates, reagent handing, pH meter maintenance, etc.
Bulb Draw: 1.8 ml
Drops/mL: 24
OAL: 15.2 cm