Learn More
Alkali Scientific Pipette Tips, 5mL, Bagged, 10 Bags of 100 Tips, 1000/cs
SDP
Catalog No. 502335764
Encompass_Preferred
Macro 5mL tips are specially designed for an ideal and comfortable fit on our Macro size pipettes. Supplied in bags, these tips are made of ultra-clear polypropylene and are guaranteed to create the most ideal seal on ProPette™ Pipettors. 1000/cs
Catalog No. 50-233-5764 Supplier Alkali Scientific Supplier No. P9305
Macro 5mL tips are specially designed for an ideal and comfortable fit on our Macro size pipettes. Supplied in bags, these tips are made of ultra-clear polypropylene and are guaranteed to create the most ideal seal on ProPette™ Pipettors. 1000/cs