Learn More
Pipette Supplies, Inc. PIPETLITE ORING 1000UL 5PK
SDP
Catalog No. 501317570
Encompass
Piston o-ring for the following Rainin 1000μL pipettes:Pipet-Lite (L, SL)Pipet-Lite XLS (XLS)Pipet-Plus (R, RL)EDP1 (E1, SE1)EDP3, EDP3 Plus (E3, SE3)E-Man Hybrid (H, SH)E4 XLS (E4, SE4)Alternative to Rainin 6200-162 / 17003377
Catalog No. 50-131-7570 Supplier Pipette Supplies, Inc. Supplier No. 21000/5
Piston o-ring for the following Rainin 1000μL pipettes:Pipet-Lite (L, SL)Pipet-Lite XLS (XLS)Pipet-Plus (R, RL)EDP1 (E1, SE1)EDP3, EDP3 Plus (E3, SE3)E-Man Hybrid (H, SH)E4 XLS (E4, SE4)Alternative to Rainin 6200-162 / 17003377