Learn More
Pipette Supplies, Inc. PIPETLITE XLS+ GREASE 1 GRAM
SDP
Catalog No. 501317580
Encompass
Used with E4 XLS+ & Pipet-LiteXLS+ pipettes up to 2000ul, and all previous pipettes being converted to the XLS+ sealing system.Identical silicone lubricant sold by Rainin. Alternative to Rainin 17014531, 6200-524
Catalog No. 50-131-7580 Supplier Pipette Supplies, Inc. Supplier No. 2XLSGR
Used with E4 XLS+ & Pipet-LiteXLS+ pipettes up to 2000ul, and all previous pipettes being converted to the XLS+ sealing system.Identical silicone lubricant sold by Rainin. Alternative to Rainin 17014531, 6200-524