Description
- Fully adjustable, air-displacement pipette with selected volume shown on digital indicator (volumeter)
- Tip holder and tip ejector are autoclavable to avoid cross-contamination
- Each pipet bears engraved identification number for traceability
Specifications
Specifications
| Accuracy | ±0.024, ±12 μL |
| Compatible Tips | D10, DL10, DF10ST, DFL10ST |
| Material | PVDF, Stainless Steel |
| Model | P2 |
| Product Line | Pipetman Classic |
| Type | Pipette |
| Volume (Metric) | 0.2 to 2 μL |
| For Use With (Application) | PCR, DNA sequencing |