Learn More
Gilson™ PIPETMAN Classic™ Pipets PROMO
Catalog No. F144802GPR Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
0.2 to 2 μL
1 to 10 μL
2 to 20 μL
20 to 100 μL
200 to 1000 μL
50 to 200 μL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
F144802GPR 1 to 10 μL
F144801GPR 0.2 to 2 μL
F123600GPR 2 to 20 μL
F123615GPR 20 to 100 μL
F123601GPR 50 to 200 μL
F123602GPR 200 to 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. F144802GPR Supplier Gilson™ Supplier No. F144802PR
Gilson™ PIPETMAN Classic™ Pipets are individually calibrated, delivered with a performance check report, and include dry seal technology for air-tightness.
  • Fully adjustable, air-displacement pipette with selected volume shown on digital indicator (volumeter)
  • Tip holder and tip ejector are autoclavable to avoid cross-contamination
  • Each pipet bears engraved identification number for traceability
Specifications
Accuracy ±0.025, ±2.5 μL
Compatible Tips D10, DL10, DF10ST, DFL10ST
Material PVDF, Stainless Steel
Model P10
Product Line Pipetman Classic
Type Pipette
Volume (Metric) 1 to 10 μL
For Use With (Application) PCR, DNA sequencing