Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ PIPETMAN™ Classic Pipette Trade-In PROMO

Catalog No. F161201TG Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
0.2 to 2 μL
1 to 10 μL
1 to 10 mL
1 to 5 mL
2 to 20 μL
20 to 100 μL
20 to 200 μL
200 to 1000 μL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
F161201TG 1 to 10 mL
F144801TG 0.2 to 2 μL
F144802TG 1 to 10 μL
F123600TG 2 to 20 μL
F123615TG 20 to 100 μL
F123601TG 20 to 200 μL
F123602TG 200 to 1000 μL
F123603TG 1 to 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. F161201TG Supplier Gilson™ Supplier No. F161201T

missing translation for 'validMainframeMsg'

Send in your used or broken pipettes and save on select Gilson models. All used brands and models qualify and a pre-postage bag will be included with the order. Refurbished pipettes will be donated to Seeding Labs, who distributes to labs and universities around the world who need it most.

  • Fully adjustable, air-displacement pipette with selected volume range
  • Volume shown on digital indicator (volumeter)
  • Autoclavable tip holder and tip ejector help to avoid cross-contamination
  • Each pipette bears engraved identification number for traceability

Specifications

Accuracy ±30, ±40, ±60 μL
Compatible Tips D10mL
Description Only Tip Ejector, and Tip Holder are Autoclavable
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Blue
Dispensing Speed Manual
Disposable Reusable
Grip Type Ergonomic
Material PVDF, Stainless Steel
Model P10mL
Number of Channels 1
Product Line Pipetman Classic
Programmability No
Sterility Non-sterile
Type Classic Pipette
Volume (Metric) 1 to 10 mL
For Use With (Application) Large volume dispensing of aqueous fluids of moderate viscosity and density
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.