Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ PIPETMANTM DIAMONDTM Tips Reload Pack
GREENER_CHOICE

Catalog No. p-7241869 Shop All Gilson Products
Change view
Click to view available options
Model:
D1000
D200
DL10
Volume (Metric):
0.1 to 20 μL
100 to 1000 μL
2 to 200 μL
Color Code:
Dark Blue
Red
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Volume (Metric) Color Code
F167012G DL10 0.1 to 20 μL Red
F167013G D200 2 to 200 μL Yellow
F167014G D1000 100 to 1000 μL Dark Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F167012G Supplier Gilson™ Supplier No. F167012
Only null left
Add to Cart
Edge
Add to Cart

PIPETMANTM DIAMONDTM Tips Reload Pack is an easy and quick way to refill an empty TIPACKTM Box. Three times smaller than a regular tips box, it is easy to store on your bench.

Accurate and Precise

  • Using PIPETMANTM DIAMONDTM Tips with your PIPETMANTM ensures maximum accuracy and precision and delivers consistent results.
  • The perfect air-tight seal prevents leaks.
  • The tips are made of pure polypropylene to deliver the correct volume and ensure complete dispensing.
  • Volume graduations make it easy to visually check the volume aspirated.

Reliable

  • GilsonTM tips are manufactured in a clean room environment ensuring they are free of detectable contaminants that can affect your results.
  • From mold to bench, our tips are guaranteed, certified and traceable to meet the most stringent demands in many applications.

Convenient Packaging

  • Using up to 70% less plastic than a regular tip box, GilsonTM's reload pack is an eco-friendly and affordable solution.
  • Its small size makes it easy to store.
  • The racks are fully autoclavable and are color-coded for easy identification and compliance with Good Laboratory Practice (GLP).

Specifications

Volume (Metric) 0.1 to 20 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Model DL10
Length (English) 1.77 in.
Length (Metric) 45 mm
Certifications/Compliance ISO 9001
For Use With (Application) Refill empty TIPACK™ boxes
Diameter (Metric) Outer .4 mm
Format Refill
Tip Style Wide Bore
Packaging Reload pack
Quantity 96 Tips/Pk.
Product Type Universal Pipette Tips
Color Code Red
Disposable Single-use
For Use With (Equipment) Gilson Pipetman
Graduated Yes
Product Line Pipetman Diamond
Material Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.