Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ PIPETMAN™ G 4-pipette Kit PROMO

Catalog No. F167360GPM Shop All Gilson Products
Change view
Click to view available options
Number of Channels:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Number of Channels
F167360GPM 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F167360GPM Supplier Gilson™ Supplier No. F167360PM

missing translation for 'validMainframeMsg'

Gilson™ PIPETMAN™ G 4-pipette Kit features a redesigned piston mechanism that reduces the force needed to aspirate and dispense liquids.

  • New piston assembly significantly reduces the force needed to aspirate and dispense liquids
  • Come with a durable stainless steel tip ejector
  • Includes three racks of PIPETMAN™ DIAMOND Tips
  • Includes four PIPETMAN™ G pipettes: P2G, P20G, P200G, and P1000G
  • Includes four Single™ pipette Holders

Specifications

Certifications/Compliance Declaration of Conformity
Compatible Tips PIPETMAN Diamond Tips
Material Polypropylene
Model Pipetman G
Number of Channels 1
Type Pipette Kit
Warranty 3 Years
Length (English) 14.37 in.
Width (English) 12.01 in.
Height (English) 4.72 in.
Weight (English) 4.37 lb.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.