Learn More
MARSHALL SCIENTIFIC Gilson PIPETMAN G Pipette Set
SDP
Catalog No. NC2888797
Encompass_Preferred
PIPETMAN G brings a new level of comfort into Gilson range with a redesigned piston mechanism that reduces the force needed to aspirate and dispense liquids. PIPETMAN G maintains the trusted durability, accuracy, and reliability of the original PIPETMAN pipettes with comfort. PIPETMAN G offers one of the lowest pipetting forces commercially available. With a redesigned piston assembly system and high quality piston grease, PIPETMAN G guarantees hours of comfortable pipetting. PIPETMAN G relies on the legendary accuracy, precision, and reliability of PIPETMAN with improved pipetting comfort. Made of PVDF and stainless steel, PIPETMAN G offers premium durability. The PIPETMAN G clip ejector allows for convenient access to serviceable parts. Provides seven pipettes covering volume transfers from 0.2 to 5000uL.
Catalog No. NC2888797 Supplier MARSHALL SCIENTIFIC Supplier No. GGPST
PIPETMAN G brings a new level of comfort into Gilson range with a redesigned piston mechanism that reduces the force needed to aspirate and dispense liquids. PIPETMAN G maintains the trusted durability, accuracy, and reliability of the original PIPETMAN pipettes with comfort. PIPETMAN G offers one of the lowest pipetting forces commercially available. With a redesigned piston assembly system and high quality piston grease, PIPETMAN G guarantees hours of comfortable pipetting. PIPETMAN G relies on the legendary accuracy, precision, and reliability of PIPETMAN with improved pipetting comfort. Made of PVDF and stainless steel, PIPETMAN G offers premium durability. The PIPETMAN G clip ejector allows for convenient access to serviceable parts. Provides seven pipettes covering volume transfers from 0.2 to 5000uL.