Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ PIPETMAN™ G Starter Kit PROMO

Catalog No. F167900GPM Shop All Gilson Products
Change view
Click to view available options
For Use With:
Manual Air Displacement Pipette
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
F167900GPM Manual Air Displacement Pipette
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F167900GPM Supplier Gilson™ Supplier No. F167900PM

missing translation for 'validMainframeMsg'

Provides greater pipetting comfort without compromising accuracy and precision. Gilson™ PIPETMAN™ G Starter Kit is an economical solution for daily pipette users with concerns for the susceptibility of Repetitive Strain Injury (RSI).

Provides greater pipetting comfort without compromising accuracy and precision. All-in-one kit is an economical solution for daily pipette users who have concerns regarding the susceptibility of Repetitive Strain Injury (RSI)

Kit Includes:

  • Three PIPETMAN G pipettes: P20G, P200G, and P1000G (equipped with stainless steel ejectors)
  • Two PIPETMAN DIAMOND Tipacks: D200 and D1000 (racks of 96 tips each)
  • One Gilson Guide to pipetting Handbook
  • Three Plastic Tip Ejectors

Specifications

For Use With Manual Air Displacement Pipette
Material Polypropylene
Product Line Pipetman
Type Starter Kit
For Use With (Application) Manual Air Displacement Pipette
Includes P20 G, P200 G, P1000 G Pipette, plus boxes of-quality, PIPETMAN Tip D200, D1000, Stainless Steel and Plastic tip ejectors, 3 Single Pipette holders and Gilson guide to pipetting
Warranty 3 Years
Length (English) 14.37 in.
Width (English) 12.01 in.
Height (English) 4.72 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.