Learn More
Description
The PIPETMAN L Micro-volume Kit offers the PIPETMAN P2L and P10L, perfect for pipetting enzymes and reagents; and a PIPETMAN P100L, suited for small-volume dispensing of aqueous fluids of moderate viscosity and density (molecular biology, immunology, etc.)
Kit Contents:
- 3 PIPETMAN L pipettes: P2L, P10L, and P100L with Gilson certificates
- 2 racks of certified-quality PIPETMAN Diaomond Tips: DL10 and D200
- 3 SINGLE™ pipette holders
- 3 Plastic tip ejectors
- 1 Gilson guide to pipetting
- 1 Two-minute inspection poster
Specifications
Specifications
| For Use With | Manual Air Displacement Pipette |
| Material | Polypropylene |
| Product Line | Pipetman |
| Type | Pipette Kit, Micro |
| For Use With (Application) | Manual Air Displacement Pipette |
| Warranty | 1 Year |
| Length (English) | 14.57 in. |
| Width (English) | 12.2 in. |
| Height (English) | 4.72 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.