Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ PIPETMAN™ L Multichannel Pipette PROMO

Catalog No. FA10036GPM Shop All Gilson Products
Change view
Click to view available options
Number of Channels:
8
12
Volume (Metric):
200 μL
300 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Number of Channels Volume (Metric)
FA10036GPM 12 200 μL
FA10035GPM 8 200 μL
FA10037GPM 8 300 μL
FA10038GPM 12 300 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. FA10036GPM Supplier Gilson™ Supplier No. FA10036GPM

missing translation for 'validMainframeMsg'

Designed for performance and comfort, PIPETMAN™ L Multichannel Pipette comes in a comprehensive range of models, adapted to your needs in the lab.

Available as a fixed or variable volume, PIPETMAN™ L air-displacement pipettes cover a full volume range, including:

  • Variable volume single channel models covering a volume range from 0.2 μL to 10 mL
  • Variable volume multichannel models covering a volume range from 0.5 μL to 1200 μL, including four models equipped with V-ring (VR) at the bottom of the tip holders for a proper fit with most tip brands, covering a volume range from 20 μL to 300 μL
  • Fixed volume single channel models covering a volume range from 1 μL to 5000 μL

Specifications

Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Yellow
Dispensing Speed Manual
Disposable Reusable
For Use With Pipette Tip 2 to 200 μL
Grip Type Ergonomic
Material Polypropylene
Model 12x200LVR
Number of Channels 12
Product Line Pipetman
Programmability No
Sterility Non-sterile
Type Pipette
Volume (Metric) 200 μL
For Use With (Application) Manual Liquid Handling
Quantity 1 Ea.
Width (English) 5.98 in.
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.