Learn More
Gilson™ PIPETMAN™ L Multichannel Pipettes PROMO
Catalog No. FA10013GPM Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
Catalog No. Volume (Metric)
FA10013GPM 0.5 to 10 μL
1 options
Catalog No. FA10013GPM Supplier Gilson™ Supplier No. FA10013GPM
New, lightweight multichannel pipette with lockable volume that has been designed to fit naturally in your hand.
  • Gilson–patented volume locking system guarantees no accidental change in volume while pipetting.
  • Adjustable tip ejector provides comfort for right- and left-handed users.
  • Redesigned handle fits naturally in your hand, no matter the size.
  • Unique internal design applies even, equal pressure from either side of the pipette, rather than on the center of the channels like other multichannels do.
  • 2D identification code tracks product reference number, date of manufacture, serial number and maximum volume of pipette.
  • Available in 8–and 12–channel covering a volume range from 0.5 to 300μL.
Specifications
Color Red
Model P8x10L
Number of Channels 8
Product Line Pipetman L
Type Pipette
Volume (Metric) 0.5 to 10 μL
Finger Style Ergonomic
Warranty 1 Year
Length (English) 5.98 in.
Width (English) 12.83 in.
Height (English) 1.54 in.
Weight (English) 0.81 lb.