Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ PIPETMANTM L Mechanical Pipette Trade-In PROMO
GREENER_CHOICE

Catalog No. fa10003mtg Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
0.2 to 2 μL
1 to 10 μL
1 to 10 mL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
500 to 5000 μL
Color:
Blue
Orange
Purple
Red
Salmon
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
FA10003MTG 2 to 20 μL Yellow
FA10001MTG 0.2 to 2 μL Orange
FA10002MTG 1 to 10 μL Red
FA10004MTG 10 to 100 μL Salmon
FA10005MTG 20 to 200 μL Yellow
FA10006MTG 100 to 1000 μL Blue
FA10007TG 500 to 5000 μL Purple
FA10008TG 1 to 10 mL Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. FA10003MTG Supplier Gilson™ Supplier No. FA10003MT
Add to Cart
Add to Cart

Send in your used or broken pipettes and save on select Gilson models. All used brands and models qualify and a pre-postage bag will be included with the order. Refurbished pipettes will be donated to Seeding Labs, who distributes to labs and universities around the world who need it most.

  • Redesigned handle fits naturally in user's hand, regardless of size
  • Volume locking system guarantees no accidental changes in volume while pipetting
  • Adjustable tip ejector provides comfort for right- and left-handed users
  • 2D identification code allows for easy tracking of product's information
  • New piston assembly designed to drastically improve pipetting and purge forces
  • Redesigned tip ejector shape and internal mechanism reduces tip ejection forces up to 50%

Specifications

Accuracy ±0.1, ±0.1, ±0.2 μL
Compatible Tips Stainless Steel Ejector
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Yellow
Dispensing Speed Manual
Disposable Reusable
For Use With D-200, DF30ST Tip
Grip Type Ergonomic
Material Polypropylene
Model P20L
Number of Channels 1
Product Line Pipetman
Programmability No
Sterility Non-sterile
Type Mechanical Pipette
Volume (Metric) 2 to 20 μL
Finger Style Ergonomic
For Use With (Application) Transporting a measured volume of liquid
For Use With (Equipment) D200, DF30ST tips
Quantity 1
Width (English) 4.25 in
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.