Learn More
MARSHALL SCIENTIFIC Gilson PIPETMAN L Pipette Set
SDP
Supplier:  MARSHALL SCIENTIFIC GLPSTMV
Encompass_Preferred
Designed for performance and comfort, PIPETMAN L comes in a comprehensive range of models adapted to your needs in the lab. The Gilson PIPETMAN L Pipette Set provides variable seven volume single channel models covering a volume range from 0.2 to 5000 uL. Comfortable, precise, accurate, and reliable, use PIPETMAN L pipettes confidently in your applications. Lightweight and balanced, PIPETMAN L combines an ergonomic design with minimized pipetting forces, giving you the ability to pipette longer without fatigue. Right-handed or left-handed? No problem. Adjust the location of the tip ejector to the most comfortable position for you. Reduce pipetting errors and avoid accidental volume changes during pipetting cycles with a true volume-locking mechanism for improved accuracy and precision. PIPETMAN L variable volume models are all fully autoclavable without disassembly for maximum convenience and protection from cross-contamination. Large v
Catalog No. NC2888812