Learn More
Pipette Supplies, Inc. PIPETMAN O-RING P10ML 5PK
SDP
Catalog No. 501317586
Encompass
10mL piston o-ring for:Gilson Pipetman Classic 10mL (P10mL)Rainin Classic 10mL (PR10ML)Gilson Pipetman G 10mL (P10mLG)Gilson Pipetman L 10mL (P10mLL)Alternative to Gilson F161813, F144503 and Rainin (Pre serial # N05803D) 6192-015, 17003373, 17003469.With seal 910ML, alternative to Gilson set F161829.Made in the USA
Catalog No. 50-131-7586 Supplier Pipette Supplies, Inc. Supplier No. 310ML/5
10mL piston o-ring for:Gilson Pipetman Classic 10mL (P10mL)Rainin Classic 10mL (PR10ML)Gilson Pipetman G 10mL (P10mLG)Gilson Pipetman L 10mL (P10mLL)Alternative to Gilson F161813, F144503 and Rainin (Pre serial # N05803D) 6192-015, 17003373, 17003469.With seal 910ML, alternative to Gilson set F161829.Made in the USA