Learn More
Pipette Supplies, Inc. PIPETMAN O-RING P2, P10 5PK
SDP
Catalog No. 501317581
Encompass
Piston o-ring for:Gilson Pipetman Classic 2μL, 10μL (P2, P10)Rainin Classic 2μL, 10μL (PR2, PR10)Gilson Pipetman Neo 2μL, 10μL (P2N, P10N)Gilson Pipetman G 2μL (P2G)Gilson Pipetman L 2μL (P2L)Alternative to Gilson F161801, F144501, F144502, Rainin 400071, 17003387, 17003441.With 90002, alternative to Gilson P2 set F144861.With 90010, alternative to Gilson P10 set F144862.Made in the USA
Catalog No. 50-131-7581 Supplier Pipette Supplies, Inc. Supplier No. 30002/5
Piston o-ring for:Gilson Pipetman Classic 2μL, 10μL (P2, P10)Rainin Classic 2μL, 10μL (PR2, PR10)Gilson Pipetman Neo 2μL, 10μL (P2N, P10N)Gilson Pipetman G 2μL (P2G)Gilson Pipetman L 2μL (P2L)Alternative to Gilson F161801, F144501, F144502, Rainin 400071, 17003387, 17003441.With 90002, alternative to Gilson P2 set F144861.With 90010, alternative to Gilson P10 set F144862.Made in the USA