Learn More
Pipette Supplies, Inc. PIPETMAN O-RING P20 5PK
SDP
Catalog No. 501317582
Encompass
Piston o-ring for:Gilson Pipetman 20μL (P20)Rainin Classic 20μL (PR20)Gilson Pipetman Fixed F2, F5, F10, F20Gilson Pipetman Neo 20μL (P20N)Gilson Pipetman M 20μL (P20M)Alternative to Rainin 400013, 17003388, 17003447, Gilson F161803, F144495.With 90020, alternative to Gilson set F144863.Made in the USA
Catalog No. 50-131-7582 Supplier Pipette Supplies, Inc. Supplier No. 30020/5
Add to Cart
Add to Cart
Piston o-ring for:Gilson Pipetman 20μL (P20)Rainin Classic 20μL (PR20)Gilson Pipetman Fixed F2, F5, F10, F20Gilson Pipetman Neo 20μL (P20N)Gilson Pipetman M 20μL (P20M)Alternative to Rainin 400013, 17003388, 17003447, Gilson F161803, F144495.With 90020, alternative to Gilson set F144863.Made in the USA