Learn More
Pipette Supplies, Inc. PIPETMAN O-RING P200 5PK
SDP
Catalog No. 501317584
Encompass
Piston o-ring for:Gilson Pipetman Classic 200μL (P200)Rainin Classic 200μL (PR200)Gilson Pipetman Fixed F200Gilson Pipetman Neo 200μL (P200N)Alternative to Gilson F161807, Rainin 400001, 17003389, 17003454.With 90200, alternative to Gilson set F144865.Made in the USA
Catalog No. 50-131-7584 Supplier Pipette Supplies, Inc. Supplier No. 30200/5
Piston o-ring for:Gilson Pipetman Classic 200μL (P200)Rainin Classic 200μL (PR200)Gilson Pipetman Fixed F200Gilson Pipetman Neo 200μL (P200N)Alternative to Gilson F161807, Rainin 400001, 17003389, 17003454.With 90200, alternative to Gilson set F144865.Made in the USA