Learn More
Pipette Supplies, Inc. Gilson Pipetman / Rainin Classic Tip Holder, P200, PR200, F200, P200N, P200G, P200L
SDP
Catalog No. NC3070042
Encompass
Tip holder P200, PR200, F200, P200N, P200G, P200L for the following single channel pipettes:Gilson Pipetman Classic 200μL, P200Rainin Classic 200μL, PR200Gilson Pipetman F Fixed F200Gilson Pipetman Neo 200μL, P200NGilson Pipetman G 200μL, P200GGilson Pipetman L 200μL, P200LAlso known as a shaft or nose cone.Alternative to Gilson F144497, F161834 & Rainin 23305, 17004987.Autoclavable.Made in the USA
Catalog No. NC3070042 Supplier Pipette Supplies, Inc. Supplier No. 60200
Tip holder P200, PR200, F200, P200N, P200G, P200L for the following single channel pipettes:Gilson Pipetman Classic 200μL, P200Rainin Classic 200μL, PR200Gilson Pipetman F Fixed F200Gilson Pipetman Neo 200μL, P200NGilson Pipetman G 200μL, P200GGilson Pipetman L 200μL, P200LAlso known as a shaft or nose cone.Alternative to Gilson F144497, F161834 & Rainin 23305, 17004987.Autoclavable.Made in the USA