Learn More
Pipette Supplies, Inc. PIPETMAN SEAL P1000 1ML, 5PK
SDP
Catalog No. 501317624
Encompass
Piston seal for:Gilson Pipetman Classic 1000μL (P1000)Gilson Pipetman Fixed F250, F300, F400, F500, F1000Gilson Pipetman Neo 1000μL (P1000N)Alternative to Gilson F123888, F144498.With 31000, alternative to Gilson set F144866.Made in the USA
Catalog No. 50-131-7624 Supplier Pipette Supplies, Inc. Supplier No. 91000/5
Piston seal for:Gilson Pipetman Classic 1000μL (P1000)Gilson Pipetman Fixed F250, F300, F400, F500, F1000Gilson Pipetman Neo 1000μL (P1000N)Alternative to Gilson F123888, F144498.With 31000, alternative to Gilson set F144866.Made in the USA