Learn More
Pipette Supplies, Inc. PIPETMAN SEAL P2 2UL, 5PK
SDP
Catalog No. 501317619
Encompass
Piston seal for:Gilson Pipetman Classic 2μL (P2)Rainin Classic 2μL (PR2)Gilson Pipetman Neo 2μL (P2N)Gilson Pipetman G 2μL (P2G)Gilson Pipetman L 2μL (P2L)Alternative to Gilson F123002, F144501, Rainin 44815, 17004731, 17004782.With 30002, alternative to Gilson set F144861.Made in the USA
Catalog No. 50-131-7619 Supplier Pipette Supplies, Inc. Supplier No. 90002/5
Piston seal for:Gilson Pipetman Classic 2μL (P2)Rainin Classic 2μL (PR2)Gilson Pipetman Neo 2μL (P2N)Gilson Pipetman G 2μL (P2G)Gilson Pipetman L 2μL (P2L)Alternative to Gilson F123002, F144501, Rainin 44815, 17004731, 17004782.With 30002, alternative to Gilson set F144861.Made in the USA