Learn More
Pipette Supplies, Inc. PIPETMAN SEAL P20 20UL 5PK
SDP
Catalog No. 501317621
Encompass
Piston seal for:Gilson Pipetman Classic 20μL (P20)Gilson Pipetman Fixed F2, F5, F10, F20Gilson Pipetman M 20μL (P20M)Gilson Pipetman Neo 20μL (P20N)Alternative to Gilson F123359, F144495.With 30020, alternative to Gilson set F144863.Made in the USA
Catalog No. 50-131-7621 Supplier Pipette Supplies, Inc. Supplier No. 90020/5
Piston seal for:Gilson Pipetman Classic 20μL (P20)Gilson Pipetman Fixed F2, F5, F10, F20Gilson Pipetman M 20μL (P20M)Gilson Pipetman Neo 20μL (P20N)Alternative to Gilson F123359, F144495.With 30020, alternative to Gilson set F144863.Made in the USA