Learn More
Pipette Supplies, Inc. PIPETMAN SHAFT P10 10UL
SDP
Catalog No. 501317599
Encompass
Tip holder / shaft for:Gilson Pipetman Classic 10μL (P10)Rainin Classic 10μL (PR10)Gilson Pipetman Neo 10μL (P10N)Gilson Pipetman G 10μL (P10G)Gilson Pipetman L 10μL (P10L)Alternative to Gilson F144502, F144819, F161831 & Rainin 44819, 17004982.Autoclavable. Made in the USA.
Catalog No. 50-131-7599 Supplier Pipette Supplies, Inc. Supplier No. 60010
Tip holder / shaft for:Gilson Pipetman Classic 10μL (P10)Rainin Classic 10μL (PR10)Gilson Pipetman Neo 10μL (P10N)Gilson Pipetman G 10μL (P10G)Gilson Pipetman L 10μL (P10L)Alternative to Gilson F144502, F144819, F161831 & Rainin 44819, 17004982.Autoclavable. Made in the USA.