Learn More
Pipette Supplies, Inc. PIPETMAN SHAFT P2 2UL
SDP
Catalog No. 501317598
Encompass
Tip holder / shaft for:Gilson Pipetman Classic 2μL (P2)Rainin Classic 2μL (PR2)Gilson Pipetman Neo 2μL (P2N)Gilson Pipetman G 2μL (P2G)Gilson Pipetman L 2μL (P2L)Alternative to Gilson F144501, F144816, F161830 & Rainin 44816, 17004985.Autoclavable. Made in the USA.
Catalog No. 50-131-7598 Supplier Pipette Supplies, Inc. Supplier No. 60002
Tip holder / shaft for:Gilson Pipetman Classic 2μL (P2)Rainin Classic 2μL (PR2)Gilson Pipetman Neo 2μL (P2N)Gilson Pipetman G 2μL (P2G)Gilson Pipetman L 2μL (P2L)Alternative to Gilson F144501, F144816, F161830 & Rainin 44816, 17004985.Autoclavable. Made in the USA.