Learn More
Pipette Supplies, Inc. PIPETMAN SHAFT P20 20UL
SDP
Catalog No. 501317600
Encompass
Tip Holder / shaft for:Gilson Pipetman Classic 20μL (P20)Rainin Classic 20μL (PR20)Gilson Pipetman F Fixed F2, F5, F10, F20Gilson Pipetman Neo 20μL (P20N)Gilson Pipetman G 20μL (P20G)Gilson Pipetman L 20μL (P20L)Alternative to Gilson F123353, F144495, F161832 & Rainin 23353, 17004986.Autoclavable.Made in the USA
Catalog No. 50-131-7600 Supplier Pipette Supplies, Inc. Supplier No. 60020
Tip Holder / shaft for:Gilson Pipetman Classic 20μL (P20)Rainin Classic 20μL (PR20)Gilson Pipetman F Fixed F2, F5, F10, F20Gilson Pipetman Neo 20μL (P20N)Gilson Pipetman G 20μL (P20G)Gilson Pipetman L 20μL (P20L)Alternative to Gilson F123353, F144495, F161832 & Rainin 23353, 17004986.Autoclavable.Made in the USA