Learn More
Pipette Supplies, Inc. PIPETMAN TIP EJECT ADAPT P2/10
SDP
Catalog No. 501317634
Encompass
Adaptable tip ejector with plastic adapter (metal and plastic) for:Gilson Pipetman Classic 2μL (P2), 10μL (P10)Gilson Pipetman Neo 2μL (P2N), 10μL (P10N)The P2 & P10 models incorporate a dual position tip ejector adapter which allow for the use of Gilson PIPETMAN D10 (short) tips or PIPETMAN DL10 (long) tips.Replaces GI F144829Made in France
Catalog No. 50-131-7634 Supplier Pipette Supplies, Inc. Supplier No. GI F144876
Adaptable tip ejector with plastic adapter (metal and plastic) for:Gilson Pipetman Classic 2μL (P2), 10μL (P10)Gilson Pipetman Neo 2μL (P2N), 10μL (P10N)The P2 & P10 models incorporate a dual position tip ejector adapter which allow for the use of Gilson PIPETMAN D10 (short) tips or PIPETMAN DL10 (long) tips.Replaces GI F144829Made in France