Learn More
Pipette Supplies, Inc. PIPETMAN ULTRA CUSTOMER CALIBR
SDP
Catalog No. NC0366316
Encompass
Pipetman Ultra Customer Calibration Tool. Gilson manufactured (red plastic/2 black prong) customer calibration tool. Note: This tool will not stop the LCD display from flashing Need Service, but it is all you need to calibrate the pipette. Gilson has a tool they use themselves which will stop the LCD display message, but that tool is not available for purchase.
Catalog No. NC0366316 Supplier Pipette Supplies, Inc. Supplier No. PS F20751
Pipetman Ultra Customer Calibration Tool. Gilson manufactured (red plastic/2 black prong) customer calibration tool. Note: This tool will not stop the LCD display from flashing Need Service, but it is all you need to calibrate the pipette. Gilson has a tool they use themselves which will stop the LCD display message, but that tool is not available for purchase.