Learn More
Gilson™ PIPETMAXTM
Catalog No. 32100001 Shop All Gilson Products
Change view
Click to view available options
Includes:
Cover Cutouts
External Safety Interlock
Standard Cover
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
32-100-001 Cover Cutouts
32-100-000 Standard Cover
32-100-002 External Safety Interlock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 32-100-001 Supplier Gilson™ Supplier No. 32100001
Add to Cart
Add to Cart
Request a Quote
PIPETMAXTM is an automated pipetting solution for the efficient processing of high-throughput biological assays, all in a benchtop size that easily fits into any lab.
PIPETMAX is built upon PIPETMANTM technology, helping to reduce costly technical replicates and maximize reproducibility by eliminating inconsistencies due to common pipetting errors. The automated system frees up time devoted to tedious pipetting jobs that improve lab productivity and allows you get to results faster. Several pre-developed, validated protocols are available for download while custom protocols can be written to accommodate any workflow.
  • Hardware and software of the PIPETMAX are customizable to fit specific needs of the assay, allowing you to configure and customize runs to any reagent, kit, or protocol
  • 8-channel cassettes can be set to use 1 to 8 tips
  • With a volume range from 1 to 1000 μL, PIPETMAX is an ideal solution for qPCR, NGS reactions, and cell-based assays
  • Small footprint and easily fits on lab bench or under a fume hood
  • Easy to move to location most suited for the assays at hand
Specifications
Product Type Digital Dispenser
Certifications/Compliance ISO 9001
Channels 8
Material Polypropylene, Glass
Volume (Metric) 1 to 1000 μL
For Use With (Application) Liquid Handling
For Use With (Equipment) 96- and 384-well microplates, strip plates, and cell culture formats
Includes Cover Cutouts
Interface Touchscreen tablet, or PC control via USB and TRILUTION™ micro software
Software TRILUTION™
Tip Material Polypropylene
Electrical Requirements 50/60 Hz, 100/240 V AC, 24 V DC, 6.25 A, 150 W
Voltage 100/240 V
Wattage 150 w
Amperage 6.25 A
Frequency 50/60 Hz