Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ PIPETMAXTM Tablet Holder

Catalog No. 32000241 Shop All Gilson Products
Change view
Click to view available options
For Use With (Equipment):
PIPETMAX™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
32-000-241 PIPETMAX™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-000-241 Supplier Gilson™ Supplier No. 32000241
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tablet holder for use with its automated pipettor.

This convenient tablet holder is for use with the GilsonTM PIPETMAXTM.

Specifications

Product Type Pipetter Accessory
Autoclavable Not Autoclavable
Color Black
Material Polypropylene
For Use With (Application) Liquid Handling
For Use With (Equipment) PIPETMAX™
Warranty 1 Year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.