Learn More
Chemglass Life Sciences Pipets, Polypropylene, Pasteur, 9", Bulk Packed, Non-Sterile, Bulk Packed, Tab Lock Box
SDP
Catalog No. 501897610
Encompass
Polypropylene Pasteur pipets offer the aspirating precision of a glass pipet and the convenience of a plastic, ready-to-use product. The virgin polypropylene pipets offer a safer alternative to glass pipets. Pipets are available in two sizes: 5-3/4¿ and 9¿ for easier handling and longer reach for working with larger vessels.
Catalog No. 50-189-7610 Supplier Chemglass Life Sciences Supplier No. CGE3489B09
Polypropylene Pasteur pipets offer the aspirating precision of a glass pipet and the convenience of a plastic, ready-to-use product. The virgin polypropylene pipets offer a safer alternative to glass pipets. Pipets are available in two sizes: 5-3/4¿ and 9¿ for easier handling and longer reach for working with larger vessels.