Learn More
Hach Company Xenosep Pipets, Transfer, PE, 15mL, 48/PK
Supplier:  Hach Company 2943400
Encompass
Xenosep Polyethylene Transfer Pipets, 15mL. Used to conveniently rinse the funnel walls with hexane extract from the sample bottle to achieve high recoveries with minimal solvent. 48 pipets per pack.
Catalog No. 50-199-6897