Learn More
CELLTREAT Scientific Products 1-5mL Pipette, Adjustable Volume, Single Channel
SDP
Catalog No. NC3377692
Encompass_Preferred
High precision adjustable pipette with easy-to-read volume displayContinuously adjustable with locking ring to prevent volume changes while pipettingLightweight design and easy operation with either handReduced operating forces; plunger is easy to press and dispenseStainless steel, PTFE and PVDF components for long lifeIt is not recommended to autoclave more than 6 timesFits CELLTREAT Scientific Products Universal Pipette Tips for accurate pipetting5mL & 10mL models do not feature tip ejection
Catalog No. NC3377692 Supplier CELLTREAT Scientific Products Supplier No. 230313
High precision adjustable pipette with easy-to-read volume displayContinuously adjustable with locking ring to prevent volume changes while pipettingLightweight design and easy operation with either handReduced operating forces; plunger is easy to press and dispenseStainless steel, PTFE and PVDF components for long lifeIt is not recommended to autoclave more than 6 timesFits CELLTREAT Scientific Products Universal Pipette Tips for accurate pipetting5mL & 10mL models do not feature tip ejection