Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Pipette Aide Holder - 6 Position
SDP

Catalog No. 01259190
Change view
Click to view available options
Mounting:
Magnet Mount
Tape Mount
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
01-259-190 Wall Mount
01-259-139 Magnet Mount
01-259-140 Tape Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
N/A
Catalog No. 01-259-190 Supplier Poltex Supplier No. MEGABRK6W
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
N/A

Holds popular brands of pipette controllers including Drummond Pipette-Aides or equivalent. Choose from Wall Mount, HB Tape Mount, or Magnet Mount with durable PETG construction.

  • Holds most popular brands of pipette controllers.
  • Holds Drummond Pipette-Aides or equivalent pipette controller.
  • Choose from Wall Mount (screws/anchors included), HB Tape Mount (high-bond tape included), or Magnet Mount (powerful magnets included).
  • Durable and cleanable PETG construction.
  • Consumables not included.

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Pipette Aid, Pipettes
Material PETG
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Application) Pipette Controller Holder
For Use With (Equipment) Pipette Controllers
Mounting Wall Mount
No. of Racks 1
No. per Case 1
No. per Pack 1
Quantity 1 Ea.
Dimensions (L x W x H) 2.25 x 10.5 x 3 in.
Weight (Metric) 1 lb.
Includes Mounting Hardware
Product Type Pipette Holder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.