Learn More
Pipette Supplies, Inc. HandyStep Electronic / AutoRep E NiMH Battery Pack
SDP
Catalog No. NC1844269
Encompass
Rechargeable battery made of four NiMH AAA cells, 4.8V/730mAh. Battery for the BrandTech HandyStep Electronic Repeating Pipette & Rainin AutoRep E Repeating Dispenser.
Catalog No. NC1844269 Supplier Pipette Supplies, Inc. Supplier No. PS 705025
Rechargeable battery made of four NiMH AAA cells, 4.8V/730mAh. Battery for the BrandTech HandyStep Electronic Repeating Pipette & Rainin AutoRep E Repeating Dispenser.