Learn More
Chemglass Life Sciences Chemglass Pipette Canister, 22.9 cm, Stainless Steel, Square
SDP
Catalog No. NC3071733
Encompass
The Chemglass pipette canister provides secure, corrosion-resistant storage and protection for laboratory pipettes. Made from 304 stainless steel and fitted with silicone pads, the square canister keeps pipettes upright and cushioned during drying or storage.
  • Constructed from 304 stainless steel for corrosion resistance.
  • 22.9 cm length with 5.1 cm (2 in) square cross-section.
  • Silicone pads on top and bottom protect pipette ends.
  • Square shape holds standard pipettes upright for drying and storage.
Catalog No. NC3071733 Supplier Chemglass Life Sciences Supplier No. CLS1710003
The Chemglass pipette canister provides secure, corrosion-resistant storage and protection for laboratory pipettes. Made from 304 stainless steel and fitted with silicone pads, the square canister keeps pipettes upright and cushioned during drying or storage.
  • Constructed from 304 stainless steel for corrosion resistance.
  • 22.9 cm length with 5.1 cm (2 in) square cross-section.
  • Silicone pads on top and bottom protect pipette ends.
  • Square shape holds standard pipettes upright for drying and storage.