Learn More
Electron Microscopy Sciences ProPette™ Pipette Controller
SDP
Catalog No. 501929487
Encompass_Preferred
This manual inline pipette controller was designed with an ergonomic grip and bulb for simple and precise operation. It is comfortable to hold and easy to use. With the "T" bar thumb lever, one can control the aspirating and dispensing speeds from serological pipettes from 1 to 100 ml in size.
To operate squeeze the bulb to create a vacuum to draw up fluid. Push the control lever backward (upward) to aspirate liquid through a pipette, or push the lever forward (downward) to dispense the liquid. A press of the central bulb on the unit blows out any remaining fluid if necessary.
Accommodates either glass or plastic serological or volumetric pipettes ranging from 1-100 ml. Includes a replaceable .45µm hydrophobic filter to protect your samples and internal components. Two additional nose cones (green, white) for easy identification keeps you organized. Polypropylene nozzle and silicone pipette adapter are autoclavable.
Catalog No. 50-192-9487 Supplier Electron Microscopy Sciences Supplier No. 70976
Add to Cart
Add to Cart
This manual inline pipette controller was designed with an ergonomic grip and bulb for simple and precise operation. It is comfortable to hold and easy to use. With the "T" bar thumb lever, one can control the aspirating and dispensing speeds from serological pipettes from 1 to 100 ml in size.
To operate squeeze the bulb to create a vacuum to draw up fluid. Push the control lever backward (upward) to aspirate liquid through a pipette, or push the lever forward (downward) to dispense the liquid. A press of the central bulb on the unit blows out any remaining fluid if necessary.
Accommodates either glass or plastic serological or volumetric pipettes ranging from 1-100 ml. Includes a replaceable .45µm hydrophobic filter to protect your samples and internal components. Two additional nose cones (green, white) for easy identification keeps you organized. Polypropylene nozzle and silicone pipette adapter are autoclavable.