Learn More
Pipette Supplies, Inc. S1 Pipet Filler, Red
SDP
Catalog No. NC1844261
Encompass_Preferred
The S1 Pipet Filler is a cordless, lightweight pipet filler that allows longer, fatigue-free pipetting for use with graduated and volumetric glass and plastic serological pipettes. Top performer. Color: Red
Catalog No. NC1844261 Supplier Pipette Supplies, Inc. Supplier No. TS 9531
The S1 Pipet Filler is a cordless, lightweight pipet filler that allows longer, fatigue-free pipetting for use with graduated and volumetric glass and plastic serological pipettes. Top performer. Color: Red