Learn More
Pipette Supplies, Inc. 1 Pipet Filler, Blue
SDP
Catalog No. NC1844260
Encompass_Preferred
The S1 Pipet Filler is a cordless, lightweight pipet filler that allows longer, fatigue-free pipetting for use with graduated and volumetric glass and plastic serological pipettes. Top performer. Color: Blue. In stock ready to ship.
Catalog No. NC1844260 Supplier Pipette Supplies, Inc. Supplier No. TS 9521
Add to Cart
Add to Cart
The S1 Pipet Filler is a cordless, lightweight pipet filler that allows longer, fatigue-free pipetting for use with graduated and volumetric glass and plastic serological pipettes. Top performer. Color: Blue. In stock ready to ship.