Learn More
Chemglass Life Sciences Pipette Controller, Manual, Green
SDP
Catalog No. 50958488
Encompass_Preferred
Pipette Controller, Manual, Green.• Lightweight- comfortable to hold, simple to use • Precise pipetting control • Use with glass or plastic volumetric and serological pipettes• Uses replaceable 3.0µm filters • Easy to maintain and clean • Sensitive lever controls filling and delivery • Complies with ISO9001, 13485 and GLP guidelinesComfortable and easy-to-use, this controller measures 8 inches long by 2 inches wide. Precise pipetting is accomplished using glass or plastic volumetric or graduated pipettes from 0.1 to 100mL.The Pipette Controller also has a blow-out control button, uses protective / replaceable syringe filter and is easy to take apart for cleaning. The filling and delivery functions area are easily controlled with a small, sensitive lever.
Catalog No. 50-958-488 Supplier Chemglass Life Sciences Supplier No. CLS347010
Pipette Controller, Manual, Green.• Lightweight- comfortable to hold, simple to use • Precise pipetting control • Use with glass or plastic volumetric and serological pipettes• Uses replaceable 3.0µm filters • Easy to maintain and clean • Sensitive lever controls filling and delivery • Complies with ISO9001, 13485 and GLP guidelinesComfortable and easy-to-use, this controller measures 8 inches long by 2 inches wide. Precise pipetting is accomplished using glass or plastic volumetric or graduated pipettes from 0.1 to 100mL.The Pipette Controller also has a blow-out control button, uses protective / replaceable syringe filter and is easy to take apart for cleaning. The filling and delivery functions area are easily controlled with a small, sensitive lever.