Learn More
Chemglass Life Sciences Pipette Filler, Manual, Quick-Release, for up to 25mL, Red
SDP
Catalog No. 501897598
Encompass
Designed for quick filling and releasing of liquid in aspirating and serological pipettes. Easy to use and easy to clean. Simply rotate the thumb wheel to aspirate or dispense accurate measurements. Press the release lever to quickly dispense all liquid from the pipettes. Resistant to acids and alkalies.
Catalog No. 50-189-7598 Supplier Chemglass Life Sciences Supplier No. CGE347252
Designed for quick filling and releasing of liquid in aspirating and serological pipettes. Easy to use and easy to clean. Simply rotate the thumb wheel to aspirate or dispense accurate measurements. Press the release lever to quickly dispense all liquid from the pipettes. Resistant to acids and alkalies.